ENA250699060 Enamine

3-(pyridin-3-yl)-1,2-oxazol-5-amine

  • MDL number MFCD07787186
  • Cas Number : 23821-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59219

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59219
Availability details for
Quantity
  • Availability
  • Add to Cart