ENA250699062 Enamine

N-[(4-chlorophenyl)methyl]-4,5-dihydro-1,3-thiazol-2-amine

  • MDL number MFCD01681683
  • Cas Number : 72239-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59221
Availability details for
Quantity
  • Availability
  • Add to Cart