ENA250699081 Enamine

4-(4-ethoxyphenyl)-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD09746482
  • Cas Number : 412308-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59255
Availability details for
Quantity
  • Availability
  • Add to Cart