ENA250699083 Enamine

3-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD01114897
  • Cas Number : 376359-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...222
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59257

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-59257
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart