ENA250699097 Enamine

3-(6-fluoro-2,3-dioxo-2,3-dihydro-1H-indol-1-yl)propanoic acid

  • MDL number MFCD09808686
  • Cas Number : 1016767-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59293
Availability details for
Quantity
  • Availability
  • Add to Cart