ENA250699101 Enamine

1-hydroxycyclopentane-1-carbonitrile

  • MDL number MFCD00043764
  • Cas Number : 5117-85-1
  • Molecular Weight : 111.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-54139

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P273,P280,P301+P310,P302+P352,P304+P340,P311,P312,P330,P361,P363,P391,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-54139
pictograms GHS06,GHS09
hstatements H301,H311,H331,H410
Availability details for
Quantity
  • Availability
  • Add to Cart