ENA250699102 Enamine

3-chloro-2-(2-ethoxyethoxy)aniline

  • MDL number MFCD08687290
  • Cas Number : 946772-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-59298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59298
Availability details for
Quantity
  • Availability
  • Add to Cart