ENA250699114 Enamine

5-hydrazinyl-2-(propan-2-yl)-1,3-oxazole-4-carbonitrile

  • MDL number MFCD03444384
  • Cas Number : 843621-90-9
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59332
Availability details for
Quantity
  • Availability
  • Add to Cart