ENA250699115 Enamine

5-hydrazinyl-2-(2-methylpropyl)-1,3-oxazole-4-carbonitrile

  • MDL number MFCD03444385
  • Cas Number : 890013-64-6
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59337
Availability details for
Quantity
  • Availability
  • Add to Cart