ENA250699116 Enamine

2-(5-amino-1H-indol-1-yl)-N,N-dimethylacetamide

  • MDL number MFCD11213952
  • Cas Number : 1094544-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59342
Availability details for
Quantity
  • Availability
  • Add to Cart