ENA250699117 Enamine

6-methyl-2-(propan-2-yl)pyrimidin-4-amine

  • MDL number MFCD12189389
  • Cas Number : 4595-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54219

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54219
Availability details for
Quantity
  • Availability
  • Add to Cart