ENA250699121 Enamine

1-acetylpiperidine-3-carboxamide

  • MDL number MFCD11643141
  • Cas Number : 15827-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59378

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59378
Availability details for
Quantity
  • Availability
  • Add to Cart