ENA250699128 Enamine

2-methyl-1-[3-(trifluoromethyl)phenyl]-4,5,6,7-tetrahydro-1H-indol-4-one

  • MDL number MFCD02707507
  • Cas Number : 327085-04-1
  • Molecular Weight : 293.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59389
Availability details for
Quantity
  • Availability
  • Add to Cart