ENA250699130 Enamine

3-methyl-5-phenylpentanoic acid

  • MDL number MFCD11172878
  • Cas Number : 2939-13-1
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59393
Availability details for
Quantity
  • Availability
  • Add to Cart