ENA250699132 Enamine

2-(2-methyl-4-nitro-1H-imidazol-1-yl)acetic acid

  • MDL number MFCD01721818
  • Cas Number : 16230-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54557

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54557
Availability details for
Quantity
  • Availability
  • Add to Cart