ENA250699140 Enamine

5-fluoro-7-methyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD06409072
  • Cas Number : 749240-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59415

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59415
Availability details for
Quantity
  • Availability
  • Add to Cart