ENA250699144 Enamine

[2-(pyridin-3-yloxy)phenyl]methanol

  • MDL number MFCD11182955
  • Cas Number : 1094325-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59424
Availability details for
Quantity
  • Availability
  • Add to Cart