ENA250699149 Enamine

3,4-diphenyl-1H-pyrazol-5-amine

  • MDL number MFCD01107800
  • Cas Number : 63633-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59431
Availability details for
Quantity
  • Availability
  • Add to Cart