ENA250699159 Enamine

4-(ethanesulfonyl)benzoic acid

  • MDL number MFCD09931970
  • Cas Number : 21571-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59447

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-59447
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart