ENA250699162 Enamine

[1-(propan-2-yl)piperidin-3-yl]methanamine

  • MDL number MFCD07186370
  • Cas Number : 875400-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59450

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59450
Availability details for
Quantity
  • Availability
  • Add to Cart