ENA250699164 Enamine

3-(1,3-dioxaindan-5-yl)-6-chloro-[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD03307622
  • Cas Number : 448190-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-59454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59454
Availability details for
Quantity
  • Availability
  • Add to Cart