ENA250699165 Enamine

4-(quinolin-8-yloxy)aniline

  • MDL number MFCD07365099
  • Cas Number : 116253-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59455

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59455
Availability details for
Quantity
  • Availability
  • Add to Cart