ENA250699170 Enamine

1,5-dimethyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD12923555
  • Cas Number : 174151-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59466
Availability details for
Quantity
  • Availability
  • Add to Cart