ENA250699174 Enamine

1-(3-ethoxyphenyl)ethan-1-amine

  • MDL number MFCD05215235
  • Cas Number : 603945-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55249

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55249
Availability details for
Quantity
  • Availability
  • Add to Cart