ENA250699178 Enamine

2-(2-oxo-2,3-dihydro-1H-indol-3-yl)acetic acid

  • MDL number MFCD09035909
  • Cas Number : 2971-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214...216
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59490
Availability details for
Quantity
  • Availability
  • Add to Cart