ENA250699180 Enamine

2,3,4,5-tetrahydro-1lambda6-benzothiepine-1,1,5-trione

  • MDL number MFCD00461295
  • Cas Number : 22710-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59493
Availability details for
Quantity
  • Availability
  • Add to Cart