ENA250699186 Enamine

4-(cyclopropylmethoxy)phenol

  • MDL number MFCD12138943
  • Cas Number : 63659-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59532
Availability details for
Quantity
  • Availability
  • Add to Cart