ENA250699187 Enamine

9-(3-chloropropyl)-9H-purin-6-amine

  • MDL number MFCD14705591
  • Cas Number : 19255-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 339...341
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55477

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55477
Availability details for
Quantity
  • Availability
  • Add to Cart