ENA250699189 Enamine

2-(2,4-dimethylphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD11104124
  • Cas Number : 1152597-25-5
  • Molecular Weight : 247.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59540
Availability details for
Quantity
  • Availability
  • Add to Cart