ENA250699215 Enamine

7-hydroxy-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD00100268
  • Cas Number : 17771-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168...170
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55846

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55846
Availability details for
Quantity
  • Availability
  • Add to Cart