ENA250699216 Enamine

2-(1H-pyrazol-5-yl)phenol

  • MDL number MFCD03093876
  • Cas Number : 34810-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55848

Safety Information

signalword Warning
safety_statement 26-36
msds https://www.enaminestore.com/catalog/EN300-55848
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart