ENA250699218 Enamine

6-chloro-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD01044385
  • Cas Number : 25369-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 298-302
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17325

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-17325
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart