ENA250699220 Enamine

ethyl 4-(2-ethoxyphenyl)-2,4-dioxobutanoate

  • MDL number MFCD12621844
  • Cas Number : 1019380-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55965
Availability details for
Quantity
  • Availability
  • Add to Cart