ENA250699221 Enamine

[3-(dimethylamino)-2-hydroxypropyl](methyl)amine

  • MDL number MFCD11135038
  • Cas Number : 34569-34-1
  • Molecular Weight : 132.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56004
Availability details for
Quantity
  • Availability
  • Add to Cart