ENA250699244 Enamine

6-bromopyridazine-3-carboxylic acid

  • MDL number MFCD06200883
  • Cas Number : 65202-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-150
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56121

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-56121
pictograms GHS06
hstatements H301,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart