ENA258389338 Enamine

2-bromo-5H-[1,3,4]thiadiazolo[3,2-a]pyrimidin-5-one

  • MDL number MFCD14705569
  • Cas Number : 1223490-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51223
Availability details for
Quantity
  • Availability
  • Add to Cart