ENA258389343 Enamine

6-chloro-3-(prop-2-en-1-yloxy)pyridine-2-carboxylic acid

  • MDL number MFCD14705580
  • Cas Number : 1221722-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54524
Availability details for
Quantity
  • Availability
  • Add to Cart