ENA258389352 Enamine

3,5-bis(2,2,2-trifluoroethoxy)aniline hydrochloride

  • MDL number MFCD14705645
  • Cas Number : 1235438-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56949

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56949
Availability details for
Quantity
  • Availability
  • Add to Cart