ENA258389363 Enamine

5-[2-(3,5-dimethoxyphenyl)ethyl]-1H-pyrazol-3-amine

  • MDL number MFCD14705758
  • Cas Number : 1000895-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59407
Availability details for
Quantity
  • Availability
  • Add to Cart