ENA258389366 Enamine

4-(aminomethyl)-N-(2-methoxyethyl)aniline

  • MDL number MFCD14705771
  • Cas Number : 1221722-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59475
Availability details for
Quantity
  • Availability
  • Add to Cart