ENA258389370 Enamine

3,3,3-trifluoro-1-(piperazin-1-yl)propan-1-one hydrochloride

  • MDL number MFCD14705777
  • Cas Number : 1221722-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59535
Availability details for
Quantity
  • Availability
  • Add to Cart