ENA258389373 Enamine

(NE)-N-[amino(dodecylsulfanyl)methylidene]-2-methoxyethan-1-amine hydrobromide

  • MDL number MFCD14705781
  • Cas Number : 1221728-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59562

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59562
Availability details for
Quantity
  • Availability
  • Add to Cart