ENA258389374 Enamine

(dodecylsulfanyl)-N'-(furan-2-ylmethyl)methanimidamide hydrobromide

  • MDL number MFCD14705782
  • Cas Number : 1221728-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59563
Availability details for
Quantity
  • Availability
  • Add to Cart