ENA258389375 Enamine

ethyl 5-acetyl-1,2,3,4-tetrahydropyridine-1-carboxylate

  • MDL number MFCD14705783
  • Cas Number : 1221723-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59569
Availability details for
Quantity
  • Availability
  • Add to Cart