ENA258389377 Enamine

2-amino-N-(2-chloro-4-methylphenyl)acetamide hydrochloride

  • MDL number MFCD14705785
  • Cas Number : 1221722-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247...249
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59573

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59573
Availability details for
Quantity
  • Availability
  • Add to Cart