ENA258389379 Enamine

ethyl 2-amino-2-(naphthalen-1-yl)acetate hydrochloride

  • MDL number MFCD14705788
  • Cas Number : 1251924-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59585
Availability details for
Quantity
  • Availability
  • Add to Cart