ENA258389380 Enamine

4-[(benzyloxy)methyl]benzene-1-carboximidamide hydrochloride

  • MDL number MFCD14705789
  • Cas Number : 1221725-98-9
  • Molecular Weight : 276.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(benzyloxy)methyl]benzenecarboximidamide hydrochloride
Inchi Code 1S/C15H16N2O.ClH/c16-15(17)14-8-6-13(7-9-14)11-18-10-12-4-2-1-3-5-12;/h1-9H,10-11H2,(H3,16,17);1H
Inchi Key NISUDWRTMUWOKG-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59586
Availability details for
Quantity
  • Availability
  • Add to Cart