ENA258389382 Enamine

4-{[(2-chlorophenyl)methyl]amino}butanoic acid hydrochloride

  • MDL number MFCD14705792
  • Cas Number : 1221723-20-1
  • Molecular Weight : 264.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59605
Availability details for
Quantity
  • Availability
  • Add to Cart