ENA258389383 Enamine

2-(5-propyl-1,2,4-oxadiazol-3-yl)propan-2-amine hydrochloride

  • MDL number MFCD14705793
  • Cas Number : 1221725-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59607
Availability details for
Quantity
  • Availability
  • Add to Cart