ENA258389384 Enamine

2-(5-tert-butyl-1,2,4-oxadiazol-3-yl)propan-2-amine hydrochloride

  • MDL number MFCD14705794
  • Cas Number : 1221722-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59608
Availability details for
Quantity
  • Availability
  • Add to Cart